missing translation for 'onlineSavingsMsg'
Learn More

Phenomenex™ Cleanert™ Snap Caps with Septa, 11 mm, No Slit

Product Code. 31744607
Change view
Click to view available options
Color:
Green
Red
Yellow
Septum Color:
Clear/Red
Red/White
White/Red
Unit Size:
Each
Pack of 100
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Septum Color unitSize
31744607 Red White/Red Each
31743389 Red Red/White Pack of 100
31742745 Green Clear/Red Pack of 100
31749127 Red Clear/Red Pack of 100
31745125 Yellow Clear/Red Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 31744607 Supplier Phenomenex™ Supplier No. AV42010
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Accessories for use with chromatography vials and in various chromatography applications.

  • Brand: Cleanert
  • Technique: Accessory
TRUSTED_SUSTAINABILITY

Specifications

Color Red
Diameter (Metric) Outer 11 mm
Material Polyethylene
Pre-slit No
Sterility Sterile
Septum PTFE/Silicone
Quantity 100 Pk.
Closure Size 11 mm
Marking Spot No
Autoclavable Autoclavable
Product Type Cap
Septum Color White/Red
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.