Learn More
Phenomenex™ Cleanert™ Crimp Caps with Septa, 20 mm, No Slit
Description
- Brand: Cleanert
- Technique: Accessory
Specifications
Specifications
| Color | Silver |
| Diameter (Metric) Outer | 20 mm |
| Material | Aluminum |
| Pre-slit | No |
| Sterility | Sterile |
| Septum | PTFE/Silicone |
| Closure Size | 18 mm |
| Marking Spot | No |
| Autoclavable | Autoclavable |
| Product Type | Cap |