Learn More
PURUS™ Critical Print Cleanroom Notebooks
Product Code. 15793609 Shop All Purus Products
Change view
Click to view available options
Color:
Blue
White
Size:
A4
A5
A6
Ruled:
Grid
Yes
Quantity:
10/Cs.
20/Cs.
40/Cs.
Unit Size:
Pack of 10
Pack of 20
Pack of 40
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Size Ruled Quantity Unit Size
15793609 White A6 Yes 40/Cs. Pack of 40
15783609 White A6 Grid 40/Cs. Pack of 40
15773609 White A5 Yes 20/Cs. Pack of 20
15763609 White A5 Grid 20/Cs. Pack of 20
15733609 White A4 Grid 10/Cs. Pack of 10
15743609 White A4 Yes 10/Cs. Pack of 10
15713609 Blue A4 Yes 10/Cs. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 15793609 Supplier PURUS™ Supplier No. PNBCPA62240 WR
  • ONLINE EXCLUSIVE PRICE
    405.13 EUR
    289.91 EUR / Pack of 40
    Save 115.22 EUR
    28% Off
Estimated Shipment: 02-02-2027
Critical Print Notebooks are ideal for writing and data recording in cleanroom environments.
For writing in cleanroom environments
  • Cleanroom processed and packaged
  • Suitable for use in ISO 4 to 6 (Class 10 to Class 1000) cleanrooms
  • Proprietary Binding Technology without organic fillers
  • Recyclable
  • College Rule
  • Amide and Silicone Free
Specifications
Material Polyethylene
Ruled Yes
Color White
Cover Material Poly
Dimensions (L x W) 148 x 105 mm
Width (Metric) 105 mm
Autoclavable Autoclavable
Certifications/Compliance Suitable for use in ISO 4 to 6 (Class 10 to Class 1000)
For Use With (Application) For writing in cleanroom environments
Product Type Cleanroom Notebook
Quantity 40/Cs.
Size A6
Weight (Metric) 80 gsm