Learn More
Abeba™ Classic 31010 Shoes
Description
- CE EN ISO 20345:2011 SB SRC
- Grained leather black
- Steel toe cap
- Slip resistance PU outsole sole
- ESD dissipative according to DIN EN 61340
- Heel strap non-foldable, adjustable
- Lining microfiber, respiration absorbing
- Adjustable instep area
- Buckle fastening
- Heel patch microfiber
Specifications
Specifications
| Certifications/Compliance | CE EN ISO 20345:2011 SB SRC |
| Color | Black |
| Liner Material | Microfiber |
| Material | Leather |
| Size | 38 |
| Sole Features | Slip-resistant |
| Sole Material | Polyurethane |
| Style | 31010 |
| Toe Material | Steel |
| Resistant To | Animal fats, oil and fuel |