Learn More
VITLAB™ Class B Polypropylene Short Form Measuring Cylinders
Product Code. 11883662
Change view
Click to view available options
Graduations:
0.5 mL
1 mL
2 mL
5 mL
10 mL
20 mL
Unit Size:
Each
Pack of 12
Pack of 6
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Graduations unitSize
11883662 5 mL Pack of 6
11803672 20 mL Pack of 6
10380221 20 mL Each
11873662 2 mL Pack of 12
11853662 0.5 mL Pack of 12
11893662 10 mL Pack of 6
11863662 1 mL Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 11883662 Supplier VITLAB™ Supplier No. 643941
Only null left
Add to basket
Add to basket
Use these cylinders for food industry and volume measurement applications. VITLAB™ Class B Polypropylene Short Form Measuring Cylinders feature easy to read, raised scale and ring marks at the primary scale points.
  • Highly transparent polypropylene graduated cylinder
  • Calibrated (In)
  • Class B tolerances according to DIN 12681 / ISO 6706.
  • Thermal stress up to 80°C does not cause tolerance limits to be permanently exceeded
Specifications
Material Polypropylene
Tolerance ±5 mL
Class B
Diameter (Metric) 51 mm
Graduations 5 mL
Limit of Error ±5mL
Shape Round
Height (Metric) 192 mm
Type Graduated Cylinder