Learn More
Hirschmann™ Class B Measuring Cylinders with NS Ground
Product Code. 11892851
Change view
Click to view available options
Capacity (Metric):
10 mL
25 mL
100 mL
250 mL
500 mL
2000 mL
Unit Size:
Each
Pack of 2
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11892851 10 mL Pack of 2
11802861 25 mL Pack of 2
11822861 100 mL Pack of 2
11832861 250 mL Pack of 2
15599025 2000 mL Each
11842861 500 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11892851 Supplier Hirschmann™ Supplier No. 2330160
  • ONLINE EXCLUSIVE PRICE
    47.00 EUR
    41.10 EUR / Pack of 2
    Save 5.90 EUR
    12% Off
Estimated Shipment: 28-12-2026
Excellent for precise liquid measurement and fluid transfer in the laboratory. Hirschmann Instruments™ Class B Measuring Cylinders with NS Ground are manufactured from durable DURAN™ glass tubing.
  • With NS ground and polyethylene stopper
  • Acc. to DIN EN ISO 4788
  • Short line, blue graduation
  • Corrosion-free and resistant to thermal shock
Specifications
Tolerance ±0.15 mL
Material Glass
Certifications/Compliance DIN EN ISO 4788
Class Class B
Capacity (Metric) 10 mL
Height (Metric) 160 mm
Includes Polyethylene Stopper
Stopper Material Polyethylene
Graduations 0.2 mL
Base Type Hexagonal
Scale Color Blue
Shape Round
Product Type Graduated Mixing Cylinder