Learn More
Hirschmann™ Class B Measuring Cylinders, Amber Stain Graduation
Product Code. 13177929
Change view
Click to view available options
Capacity (Metric):
5 mL
10 mL
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
5000 mL
Unit Size:
Each
Pack of 2
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
13177929 100 mL Pack of 2
10029000 1000 mL Each
11869201 10 mL Pack of 2
10039000 2000 mL Each
12562736 25 mL Pack of 2
10537641 5000 mL Each
10144011 50 mL Pack of 2
10626525 5 mL Pack of 2
10481862 500 mL Pack of 2
10779012 250 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 13177929 Supplier Hirschmann™ Supplier No. 2200180
  • ONLINE EXCLUSIVE PRICE
    30.00 EUR
    26.10 EUR / Pack of 2
    Save 3.90 EUR
    13% Off
Estimated Shipment: 14-12-2026
Excellent for precise liquid measurement and fluid transfer in the laboratory. Hirschmann Instruments™ Class B Measuring Cylinders, Amber Stain Graduation are manufactured from durable DURAN™ glass tubing.
  • With spout, short line
  • Amber stain graduation
  • Acc. to DIN EN ISO 4788
  • Corrosion-free and resistant to thermal shock
Specifications
Material Glass
Capacity (Metric) 100 mL
Graduations 1 mL
Limit of Error 0.75
Base Type Hexagonal
Scale Color Brown
Shape Round
Class Class B
Height (Metric) 260 mm
Certifications/Compliance DIN EN ISO 4788
Tolerance ±0.75 mL
Product Type Graduated Measuring Cylinder