Learn More
Hirschmann™ Class B Measuring Cylinders, Blue Stain Graduation
Product Code. 11788021
Change view
Click to view available options
Capacity (Metric):
5 mL
10 mL
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
Unit Size:
Each
Pack of 2
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11788021 500 mL Pack of 2
11738021 10 mL Pack of 2
11748021 25 mL Pack of 2
11812861 50 mL Pack of 2
11758021 50 mL Pack of 2
11768021 100 mL Pack of 2
11778021 250 mL Pack of 2
10573661 1000 mL Each
10668051 2000 mL Each
11796482 5 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 11788021 Supplier Hirschmann™ Supplier No. 2230190
  • ONLINE EXCLUSIVE PRICE
    83.00 EUR
    73.40 EUR / Pack of 2
    Save 9.60 EUR
    11% Off
View all promotions
Estimated Shipment: 07-10-2026
Add to basket
Add to basket
Excellent for precise liquid measurement and fluid transfer in the laboratory. Hirschmann Instruments™ Class B Measuring Cylinders, Blue Stain Graduation are manufactured from durable DURAN™ glass tubing.
  • With spout, short line
  • Blue stain graduation
  • Acc. to DIN EN ISO 4788
  • Corrosion-free and resistant to thermal shock
Specifications
Material Glass
Capacity (Metric) 500 mL
Graduations 5 mL
Limit of Error 3.75
Base Type Hexagonal
Scale Color Blue
Shape Round
Class Class B
Height (Metric) 390 mm
Certifications/Compliance DIN EN ISO 4788
Tolerance ±3.75 mL
Product Type Graduated Measuring Cylinder