Learn More
Hirschmann™ AR™ Glass Volumetric Pipettes
Product Code. 11787971
Change view
Click to view available options
Volume (Metric):
0.5 mL
1 mL
2 mL
3 mL
4 mL
5 mL
10 mL
15 mL
20 mL
25 mL
50 mL
100 mL
Unit Size:
Pack of 12
Pack of 6
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11787971 10 mL Pack of 6
11732252 0.5 mL Pack of 12
11752252 100 mL Pack of 6
11762252 15 mL Pack of 6
11757971 1 mL Pack of 12
11797971 20 mL Pack of 6
11707981 25 mL Pack of 6
11767971 2 mL Pack of 12
11742252 3 mL Pack of 6
11776652 4 mL Pack of 6
11717981 50 mL Pack of 6
11777971 5 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 11787971 Supplier Hirschmann™ Supplier No. 1300160
  • ONLINE EXCLUSIVE PRICE
    57.49 EUR
    51.35 EUR / Pack of 6
    Save 6.14 EUR
    10% Off
Estimated Shipment: 08-02-2027
Provides an extremely high degree of volumetric accuracy for your pipetting needs. Hirschmann Instruments™ AR™ Glass Volumetric Pipettes have a tolerance better than Class B.
  • One mark
  • Amber stain graduation
  • Acc. to DIN EN ISO 648
Specifications
Class B
Material Glass
Graduated Yes
Graduation Color Brown
Quantity 6/Pk.
Tolerance 0.03 mL
Volume (Metric) 10 mL
Length (Metric) 450 mm
Type Volumetric Pipet