Learn More
Hirschmann™ AR™ Glass Volumetric Pipettes, Class AS
Product Code. 11700794
Change view
Click to view available options
Volume (Metric):
1 mL
2 mL
2.5 mL
3 mL
5 mL
10 mL
15 mL
20 mL
25 mL
50 mL
100 mL
Unit Size:
Pack of 12
Pack of 6
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11700794 3 mL Pack of 6
11770784 1 mL Pack of 12
11780784 2 mL Pack of 12
11790784 2.5 mL Pack of 6
11710794 5 mL Pack of 6
11720794 10 mL Pack of 6
11770804 15 mL Pack of 6
11780804 20 mL Pack of 6
11790804 25 mL Pack of 6
11700814 50 mL Pack of 6
11710814 100 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 11700794 Supplier Hirschmann™ Supplier No. 1341148
  • ONLINE EXCLUSIVE PRICE
    51.00 EUR
    45.30 EUR / Pack of 6
    Save 5.70 EUR
    11% Off
Estimated Shipment: 16-11-2026
Provides an extremely high degree of volumetric accuracy for your pipetting needs. Hirschmann Instruments™ AR™ Glass Volumetric Pipettes, Class AS come with blue graduation.
  • Acc. to DIN EN ISO 648
  • One mark, blue graduation
  • Conformity certified, with dated batch identification
Specifications
Class AS
Material AR-GLAS
Graduated Yes
Graduation Color Blue
Quantity 6/Pk.
Tolerance 0.01 mL
Volume (Metric) 3 mL
Length (Metric) 350 mm
Type Volumetric Pipet