Learn More
Hirschmann™ Class A Volumetric Pipettes with Amber Stain Graduation, USP
Product Code. 15685455
Change view
Click to view available options
Volume (Metric):
4 mL
6 mL
7 mL
10 mL
15 mL
Tolerance:
0.001 mL
0.01 mL
0.02 mL
0.03 mL
Length (Metric):
400 mm
419 mm
440 mm
500 mm
Unit Size:
Pack of 6
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Tolerance Length (Metric) Unit Size
15685455 6 mL 0.01 mL 400 mm Pack of 6
11731526 10 mL 0.02 mL 440 mm Pack of 6
15665455 4 mL 0.001 mL 400 mm Pack of 6
12851783 15 mL 0.03 mL 500 mm Pack of 6
15605465 7 mL 0.01 mL 419 mm Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 15685455 Supplier Hirschmann™ Supplier No. 134015627
  • ONLINE EXCLUSIVE PRICE
    77.00 EUR
    68.10 EUR / Pack of 6
    Save 8.90 EUR
    11% Off
Estimated Shipment: 09-11-2026
Provides an extremely high degree of volumetric accuracy according to USP standards for your pipetting needs. Hirschmann™ Class A Volumetric Pipettes with Amber Stain Graduation are manufactured from clear AR-Glas™.
  • Acc. to USP
  • One mark
  • With dated batch identification
Specifications
Class A
Material Soda Lime Glass
Graduated Yes
Graduation Color Brown
Quantity 6/Pk.
Tolerance 0.01 mL
Volume (Metric) 6 mL
Length (Metric) 400 mm
Type Measuring Pipet