Learn More
Hirschmann™ Class A Measuring Cylinders, Amber Graduation
Product Code. 10479662
Change view
Click to view available options
Capacity (Metric):
5 mL
10 mL
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
Unit Size:
Each
Pack of 2
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10479662 250 mL Pack of 2
10542404 5 mL Pack of 2
13117939 10 mL Pack of 2
10117411 25 mL Pack of 2
12751528 50 mL Pack of 2
10505921 100 mL Pack of 2
11834382 500 mL Pack of 2
11874412 1000 mL Each
10262431 2000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 10479662 Supplier Hirschmann™ Supplier No. 2210186
  • ONLINE EXCLUSIVE PRICE
    66.00 EUR
    57.90 EUR / Pack of 2
    Save 8.10 EUR
    12% Off
Estimated Shipment: 28-12-2026
Excellent for precise liquid measurement and fluid transfer in the laboratory. Hirschmann Instruments™ Class A Measuring Cylinders, Amber Graduation are manufactured from durable DURAN™ glass tubing.
  • With spout, main point ring amber stain graduation
  • Conformity certified, with dated batch identification
  • Acc. to DIN EN ISO 4788
  • Corrosion-free and resistant to thermal shock
Specifications
Material Glass
Capacity (Metric) 250 mL
Graduations 2 mL
Limit of Error 1
Base Type Hexagonal
Scale Color Brown
Shape Round
Class Class A
Height (Metric) 335 mm
Certifications/Compliance DIN EN ISO 4788
Tolerance ±1 mL
Product Type Graduated Measuring Cylinder