Learn More
Hirschmann™ Class A Brown Graduated USP Measuring Pipettes
Product Code. 15645465
Change view
Click to view available options
Volume (Metric):
1 mL
2 mL
3 mL
8 mL
9 mL
25 mL
50 mL
100 mL
Unit Size:
Pack of 12
Pack of 6
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
15645465 9 mL Pack of 6
11711526 1 mL Pack of 12
12801783 2 mL Pack of 12
15645455 3 mL Pack of 6
15625465 8 mL Pack of 6
11741526 25 mL Pack of 6
11751526 50 mL Pack of 6
11731536 100 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 15645465 Supplier Hirschmann™ Supplier No. 134015927
  • ONLINE EXCLUSIVE PRICE
    77.00 EUR
    68.10 EUR / Pack of 6
    Save 8.90 EUR
    11% Off
Estimated Shipment: 23-11-2026
Provides an extremely high degree of volumetric accuracy for your pipetting needs.
  • With dated batch identification
  • Manufactured from AR glass™ soda-lime glass with a smooth, non-porous surface
  • With volume mark
  • Brown graduation
Specifications
Class A
Material AR-GLAS
For Use With (Application) Volumetric accuracy in pipetting
Graduated Yes
Graduation Color Brown
Tolerance 0.02 mL
Volume (Metric) 9 mL
Length (Metric) 430 ±10 mm