Learn More
Phenomenex™ Claricep™ Empty Flash Columns, Full Set with 2 Frits
Product Code. 31742277 Shop All Phenomenex Products
Change view
Click to view available options
Bed Weight (Metric):
3 kg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Bed Weight (Metric) Unit Size
31742277 3 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31742277 Supplier Phenomenex™ Supplier No. FCH5000I
Estimated Shipment: 19-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Available in two flash media (irregular and spherical) with multiple chemistries, Claricep columns are optimized for resolution and high loading capacity. The deactivated silica surface promotes compound stability, excellent peak shape and high performance for both acidic and basic compounds.
  • Brand: Claricep
  • Technique: Accessory
TRUSTED_SUSTAINABILITY
Specifications
Type Empty Cartridge
Bed Weight (Metric) 3 kg