Learn More
Phenomenex™ Claricep™ CS Silica Flash Columns, Irregular Media, Particle Size 40 to 60 μm
Product Code. 31746850 Shop All Phenomenex Products
Change view
Click to view available options
Bed Weight (Metric):
80 g
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Bed Weight (Metric) Unit Size
31746850 80 g Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31746850 Supplier Phenomenex™ Supplier No. CS1200800
 Restricted Item
Estimated Shipment: 01-12-2026
Available in two flash media (irregular and spherical) with multiple chemistries, Claricep columns are optimized for resolution and high loading capacity. The deactivated silica surface promotes compound stability, excellent peak shape and high performance for both acidic and basic compounds.
  • Brand: Claricep
  • Technique: FLASH
  • Particle Type: Irregular
TRUSTED_SUSTAINABILITY
Specifications
Type Flash Column
Bed Weight (Metric) 80 g
Quantity 5 Pk.