missing translation for 'onlineSavingsMsg'
Learn More

CKIP-1 Antibody [Alexa Fluor™ 532], Novus Biologicals™

Product Code. 30750669 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30750669 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30750669 Supplier Novus Biologicals Supplier No. NBP347115AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CKIP-1 Polyclonal antibody specifically detects CKIP-1 in Human,Mouse samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CKIP-1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias 2810052M02Rik, Casein kinase 2-interacting protein 1, C-Jun-binding protein, CK2 interacting protein 1; HQ0024c protein, CK2-interacting protein 1, CKIP1, CKIP-1PH domain-containing family O member 1, JBP, OC120, Osteoclast maturation-associated gene 120 protein, pleckstrin homology domain containing, family O member 1, pleckstrin homology domain-containing family O member 1
Host Species Rabbit
Immunogen pleckstrin homology domain containing, family O member 1. (Uniprot# Q53GL0)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 51177
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.