missing translation for 'onlineSavingsMsg'
Learn More

CJ-023423, TRC

Product Code. 30008327
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008327 1 mg 1mg
30007951 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30008327 Supplier Toronto Research Chemicals Supplier No. TRCC5358001MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 415903-37-6
Molecular Formula C26H29N5O3S
Molecular Weight (g/mol) 491.61
IUPAC Name 1-[2-[4-(2-ethyl-4,6-dimethylimidazo[4,5-c]pyridin-1-yl)phenyl]ethyl]-3-(4-methylphenyl)sulfonylurea
SMILES CCc1nc2c(C)nc(C)cc2n1c3ccc(CCNC(=O)NS(=O)(=O)c4ccc(C)cc4)cc3

Specifications

Quantity 1 mg
Synonym CJ-023423, AAT 007, CJ 023423, Grapiprant, MR 10A7, N-[[[2-[4-(2-Ethyl-4,6-dimethyl-1H-imidazo[4,5-c]pyridin-1-yl)phenyl]ethyl]amino]carbonyl]-4-methylbenzenesulfonamide, RQ 00000007, RQ 7, N-[[[2-[4-(2-Ethyl-4,6-dimethyl-1H-imidazo[4,5-c]pyridin-1-yl)phenyl]ethyl]amino]carbonyl]-4-methyl-benzenesulfonamide
InChI Formula InChI=1S/C26H29N5O3S/c1-5-24-29-25-19(4)28-18(3)16-23(25)31(24)21-10-8-20(9-11-21)14-15-27-26(32)30-35(33,34)22-12-6-17(2)7-13-22/h6-13,16H,5,14-15H2,1-4H3,(H2,27,30,32)
Chemical Name or Material 2-Ethyl-4,6-dimethyl-1-[4-[2-[(([(4-methylphenyl)sulfonyl]amino)carbonyl)amino]ethyl]phenyl]-1H-imidazo[4,5-c]pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.