missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citric Acid Monohydrate, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Citric acid monohydrate |
| CAS | 5949-29-1 |
| Analyte or Component Names | Citric acid monohydrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C6 H8 O7 . H2 O |
| Certifications/Compliance | ISO 9001 |
| Synonym | Citric Acid Monohydrate, 2-Hydroxypropane-1,2,3-tricarboxylic acid monohydrate |
| InChI Formula | InChI=1S/C6H8O7.H2O/c7-3(8)1-6(13,5(11)12)2-4(9)10;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);1H2 |
| Solution Type | Neat |
| SMILES | O.OC(=O)CC(O)(CC(=O)O)C(=O)O |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?