Learn More
Citric Acid Monohydrate, For Analysis, ACS, EMSURE™, MilliporeSigma™
Product Code. 15873840
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Plastic Bottle
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15873840 500 g Plastic Bottle 500g
15883840 1 kg Plastic Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15873840 Supplier Supelco Supplier No. 1002440500
  • ONLINE EXCLUSIVE PRICE
    75.40 EUR
    55.68 EUR / 500g
    Save 19.72 EUR
    26% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Chemical Identifiers
CAS 5949-29-1
Molecular Formula C6H10O8
Molecular Weight (g/mol) 210.138
InChI Key YASYEJJMZJALEJ-UHFFFAOYSA-N
Synonym citric acid monohydrate, citric acid hydrate, 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate, citric acid, monohydrate, unii-2968phw8qp, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate, citrate, acidum citricum monohydricum, citric acid monohydrate usp
PubChem CID 22230
ChEBI CHEBI:31404
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate
SMILES C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O
Specifications
pH 1.85
Melting Point 135°C to 152°C
Color White
Density 1.54 g/3 (20 °C)
Quantity 500 g
Packaging Plastic Bottle
Physical Form Solid
Vapor Pressure 0.1 hPa at 20°C
Solubility Information 880g/L
Formula Weight 210.14
Percent Purity 99.5 to 100.5 %
Grade ACS/ISO/Reagent Ph. Eur.
Show More Show Less
compliance-icons
Product Identifier
  • Citric acid monohydrate for analysis EMSURE ACS,ISO,Reag. Ph Eur
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash skin thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: citric acid monohydrate