missing translation for 'onlineSavingsMsg'
Learn More

Cisplatin, TRC

Product Code. 18032740
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18032740 500 mg 500mg
18012840 50 mg 50mg
18092730 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18032740 Supplier Toronto Research Chemicals Supplier No. TRCC499500500MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15663-27-1
Molecular Formula Cl2 H6 N2 Pt
Molecular Weight (g/mol) 300.04
Synonym Cisplatin, cis-Diamminedichloroplatinum(II)
IUPAC Name azanide;dichloroplatinum(2+)
SMILES [Cl-][Pt+2]([Cl-])([NH3])[NH3]

Specifications

Quantity 500 mg
InChI Formula InChI=1S/2ClH.2H3N.Pt/h2*1H;2*1H3;/q;;;;+2/p-2
Formula Weight 298.9556
Chemical Name or Material Cisplatin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.