Learn More
Cisplatin, TRC
Product Code. 18012840
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18012840 50 mg 50mg
18092730 100 mg 100mg
18032740 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 18012840 Supplier Toronto Research Chemicals Supplier No. TRCC49950050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 15663-27-1
Molecular Formula Cl2 H6 N2 Pt
Molecular Weight (g/mol) 300.04
Synonym Cisplatin, cis-Diamminedichloroplatinum(II)
IUPAC Name azanide;dichloroplatinum(2+)
SMILES [Cl-][Pt+2]([Cl-])([NH3])[NH3]
Specifications
Quantity 50 mg
InChI Formula InChI=1S/2ClH.2H3N.Pt/h2*1H;2*1H3;/q;;;;+2/p-2
Formula Weight 298.9556
Chemical Name or Material Cisplatin