missing translation for 'onlineSavingsMsg'
Learn More

cis(Z)-Flupenthixol dihydrochloride, MP Biomedicals™

Product Code. 11403072 Shop All MP Biomedicals Products
Change view
Click to view available options
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11403072 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11403072 Supplier MP Biomedicals Supplier No. 0215370280

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Antipsychotic; neuroleptic agent; dopamine receptor antagonist. Soluble in water (17mg/mL)

TRUSTED_SUSTAINABILITY

Specifications

CAS 2709-56-0
Color White
Molecular Formula C23H25F3N2OS
MDL Number MFCD00069278
Quantity 10 mg
Synonym flupentixol, cis-flupentixol, cis-z-flupenthixol, emergil, flupenthixol, siplaril, siplarol, alpha-flupenthixol, cis-flupenthixol, flupentixolum
InChI Key NJMYODHXAKYRHW-DVZOWYKESA-N
SMILES C1CN(CCN1CCC=C2C3=CC=CC=C3SC4=C2C=C(C=C4)C(F)(F)F)CCO
IUPAC Name 2-[4-[(3Z)-3-[2-(trifluoromethyl)thioxanthen-9-ylidene]propyl]piperazin-1-yl]ethanol
Molecular Weight (g/mol) 434.521
PubChem CID 5281881
ChEBI CHEBI:10454
Formula Weight 507.44
Physical Form Crystalline Powder
Chemical Name or Material cis(Z)-Flupenthixol dihydrochloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.