missing translation for 'onlineSavingsMsg'
Learn More

cis-N-Acetyl-S-(4-hydroxy-2-buten-1-yl)-L-cysteine-d3, TRC

Product Code. 30324250
Change view
Click to view available options
Quantity:
1 mg
10 mg
Product Code. Quantity
30324250 10 mg
30324816 1 mg
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30324250

Brand: Toronto Research Chemicals TRCA17864710MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material cis-N-Acetyl-S-(4-hydroxy-2-buten-1-yl)-L-cysteine-d3
Molecular Formula C9H12D3NO4S
Quantity 10 mg
Synonym cis-N-Acetyl-S-(4-hydroxy-2-butenyl)-L-cysteine-d3
InChI Formula InChI=1S/C9H15NO4S/c1-7(12)10-8(9(13)14)6-15-5-3-2-4-11/h2-3,8,11H,4-6H2,1H3,(H,10,12)(H,13,14)/b3-2-/t8-/m0/s1/i1D3
SMILES OC/C=C\CSC[C@H](NC(C([2H])([2H])[2H])=O)C(O)=O
IUPAC Name (Z)-N-(acetyl-d3)-S-(4-hydroxybut-2-en-1-yl)-L-cysteine
Molecular Weight (g/mol) 236.3
Formula Weight 236.09
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.