missing translation for 'onlineSavingsMsg'
Learn More

cis-Diamminedinitratoplatinum, TRC

Product Code. 30365317
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30365317 5 mg 5mg
30354804 50 mg 50mg
30351550 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30365317 Supplier Toronto Research Chemicals Supplier No. TRCD4162635MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41575-87-5
Molecular Formula H6N4O6Pt
Molecular Weight (g/mol) 353.15
Synonym (SP-4-2)-Diamminebis(nitrato-κO)platinum, cis-Diamminebis(nitrito-O)platinum
IUPAC Name azane;platinum(2+);dinitrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].N.N.[Pt+2]

Specifications

Quantity 5 mg
InChI Formula InChI=1S/2NO3.2H3N.Pt/c2*2-1(3)4;;;/h;;2*1H3;/q2*-1;;;+2
Formula Weight 352.9935
Chemical Name or Material cis-Diamminedinitratoplatinum
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.