missing translation for 'onlineSavingsMsg'
Learn More

cis-9-Octadecen-1-ol, Sigma-Aldrich™

Product Code. 15831261
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Unit Size:
1 L
100mL
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15831261 2.5 L 2.5 L
15811261 100 mL 100mL
15821261 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15831261 Supplier Sigma Aldrich Supplier No. 8209232500
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143-28-2

Specifications

Melting Point 2°C to 12°C
Density 0.85 g/cm3 at 20°C
Boiling Point 330°C to 360°C/1013 hPa
Molecular Formula CH3(CH2)7CH=CH(CH2)8OH
MDL Number MFCD00002993
Quantity 2.5 L
Synonym cis-9-Octadecen-1-ol, Oleyl alcohol, cis-9-Octadecen-1-ol
Molecular Weight (g/mol) 268.48
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.