Learn More
cis-1,3-O-Benzylideneglycerol, TRC
Specifications
Specifications
| Chemical Name or Material | cis-1,3-O-Benzylideneglycerol |
| CAS | 4141-19-9 |
| Molecular Formula | C10 H12 O3 |
| Quantity | 25 g |
| Synonym | 1,3-Dioxan-5-ol, 2-phenyl-, cis-, m-Dioxan-5-ol, 2-phenyl-, cis- (8CI), cis-2-Phenyl-1,3-dioxan-5-ol, cis-1,3-O-Benzylideneglycerol, cis-5-Hydroxy-2-phenyl-1,3-dioxane |
| InChI Formula | InChI=1S/C10H12O3/c11-9-6-12-10(13-7-9)8-4-2-1-3-5-8/h1-5,9-11H,6-7H2/t9-,10+ |
| SMILES | O[C@@H]1CO[C@@H](OC1)c2ccccc2 |
| IUPAC Name | 2-phenyl-1,3-dioxan-5-ol |
| Molecular Weight (g/mol) | 180.2 |
| Formula Weight | 180.0786 |