missing translation for 'onlineSavingsMsg'
Learn More

Ciproxifan maleate, MedChemExpress

Product Code. p-200101138
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16472527 50 mg 50mg
16416428 10 mg 10mg
16409017 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16472527 Supplier Medchem Express Supplier No. HY15289
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Ciproxifan maleate (FUB 359 maleate) is a potent, selective, orally bioavailable and competitive antagonist of histamine H3-receptor, with an IC50 of 9.2 nM. Ciproxifan maleate displays low apparent affinity at other receptor subtypes. Ciproxifan maleate can be used for the research of aging disorders and Alzheimer's disease.

Ciproxifan maleate (FUB 359 maleate) is a potent, selective, orally bioavailable and competitive antagonist of histamine H3-receptor, with an IC50 of 9.2 nM. Ciproxifan maleate displays low apparent affinity at other receptor subtypes. Ciproxifan maleate can be used for the research of aging disorders and Alzheimer's disease.

Specifications

Chemical Name or Material Ciproxifan maleate
CAS 184025-19-2
Color White
Purity Grade Notes Research
Molecular Formula C20H22N2O6
Quantity 50 mg
Synonym FUB 359 maleate
Solubility Information DMSO : ≥ 100 mg/mL (258.80 mM) ∣H2O : 3.57 mg/mL (9.24 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(C1CC1)C2=CC=C(OCCCC3=CNC=N3)C=C2.O=C(O)/C=C\C(O)=O
Molecular Weight (g/mol) 386.4
Formula Weight 386.4
Percent Purity 99.49%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.