missing translation for 'onlineSavingsMsg'
Learn More

CIP29 Antibody, Novus Biologicals™

Product Code. 18675882 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18675882 100 μg 100µL
18691974 50 μg 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18675882 Supplier Novus Biologicals Supplier No. NBP297653100ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CIP29 Polyclonal antibody specifically detects CIP29 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CIP29
Applications Western Blot, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunoprecipitation 1-5 μL/mg of lysate, Immunohistochemistry-Paraffin 1:50-1:200
Formulation PBS (pH 7.0)
Gene Alias CIP29Hcc-1, cytokine induced protein 29 kDa, Cytokine-induced protein of 29 kDa, HCC1MGC14726, hepatocellular carcinoma 1, HSPC316, Nuclear protein Hcc-1, proliferation associated cytokine-inducible protein CIP29, Proliferation-associated cytokine-inducible protein CIP29, SAP domain containing ribonucleoprotein, SAP domain-containing ribonucleoprotein, THO1
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human CIP29 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84324
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.