Learn More
Cinromide, TRC
Product Code. 30363664
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30363664 10 g 10g
30362935 1 g 1g
30361639 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30363664 Supplier Toronto Research Chemicals Supplier No. TRCC59646310G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 58473-74-8
Molecular Formula C11H12BrNO
Molecular Weight (g/mol) 254.12
Synonym (2E)-3-(3-Bromophenyl)-N-ethyl-2-propenamide, (E)-3-(3-Bromophenyl)-N-ethyl-2-propenamide, BW 122U, Vumide
IUPAC Name (E)-3-(3-bromophenyl)-N-ethylprop-2-enamide
SMILES CCNC(=O)\C=C\c1cccc(Br)c1
Specifications
Quantity 10 g
InChI Formula InChI=1S/C11H12BrNO/c1-2-13-11(14)7-6-9-4-3-5-10(12)8-9/h3-8H,2H2,1H3,(H,13,14)/b7-6+
Formula Weight 253.0102
Chemical Name or Material Cinromide