missing translation for 'onlineSavingsMsg'
Learn More

Cinnarizine, TRC

Product Code. 17971965
Change view
Click to view available options
Quantity:
100 mg
5 g
10 g
Unit Size:
100mg
10g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17971965 10 g 10g
17934085 100 mg 100mg
17902565 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17971965 Supplier Toronto Research Chemicals Supplier No. TRCC46530010G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 298-57-7
Molecular Formula C26 H28 N2
Molecular Weight (g/mol) 368.51
Synonym Cinnarizine, (E)-1-(Diphenylmethyl)-4-(3-phenylprop-2-enyl)piperazine, Piperazine, 1-(diphenylmethyl)-4-(3-phenyl-2-propenyl)- (9CI), Piperazine, 1-cinnamyl-4-(diphenylmethyl)- (6CI,7CI,8CI), 1-(3-Phenylallyl)-4-(diphenylmethyl)piperazine, 1-Benzhydryl-4-cinnamylpiperazine, 1-Cinnamyl-4-(diphenylmethyl)piperazine, 1-Cinnamyl-4-benzhydrylpiperazine, 1-Diphenylmethyl-4-cinnamoylpiperazine, 516MD, Aplactan, Aplexal, Apotomin, Artate, Carecin, Cerebolan, Cerepar, Cinaperazine, Cinarizine, Cinazyn, Cinnacet, Cinnageron, Cinnarizine, Cinnipirine, Corathiem, Denapol, Dimitron, Dimitronal, Eglen, Folcodal, Giganten, Glanil, Hilactan, Ixterol, Katoseran, Labyrin, Lazeta, Marisan, Midronal, Mitronal, N-Benzhydryl-N'-cinnamylpiperazine, Olamin, Processine, R 1575, R 516, Sedatromin, Sepan, Siptazin, Spaderizine, Stugeron, Stunarone, Stutgeron, Stutgin, Toliman, Vergo
IUPAC Name 1-benzhydryl-4-[(E)-3-phenylprop-2-enyl]piperazine
SMILES C(\C=C\c1ccccc1)N2CCN(CC2)C(c3ccccc3)c4ccccc4

Specifications

Quantity 10 g
InChI Formula InChI=1S/C26H28N2/c1-4-11-23(12-5-1)13-10-18-27-19-21-28(22-20-27)26(24-14-6-2-7-15-24)25-16-8-3-9-17-25/h1-17,26H,18-22H2/b13-10+
Formula Weight 368.2252
Percent Purity >95%
Chemical Name or Material Cinnarizine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.