missing translation for 'onlineSavingsMsg'
Learn More

Cinnarizine, Thermo Scientific™

An antihistamine

90.90 EUR - 236.00 EUR

Chemical Identifiers

CAS 298-57-7
Molecular Formula C26H28N2
Molecular Weight (g/mol) 368.524
MDL Number MFCD00056037
InChI Key DERZBLKQOCDDDZ-JLHYYAGUSA-N
Synonym cinnarizine, stugeron, dimitronal, stutgin, dimitron, folcodal, midronal, mitronal, stutgeron, glanil
PubChem CID 1547484
ChEBI CHEBI:31403
IUPAC Name 1-benzhydryl-4-[(E)-3-phenylprop-2-enyl]piperazine
SMILES C1CN(CCN1CC=CC2=CC=CC=C2)C(C3=CC=CC=C3)C4=CC=CC=C4
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15486459
SDS View Documents
Thermo Scientific Alfa Aesar
J64568.14
25g
90.90 EUR
25g
View alternative products
Save Up to 
»
Estimated Shipment: 25-08-2026
Add to basket
 
 Restricted Item
15496459
SDS View Documents
Thermo Scientific Alfa Aesar
J64568.22
100g
236.00 EUR
100g
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

298-57-7
368.524
DERZBLKQOCDDDZ-JLHYYAGUSA-N
1547484
1-benzhydryl-4-[(E)-3-phenylprop-2-enyl]piperazine
C26H28N2
MFCD00056037
cinnarizine, stugeron, dimitronal, stutgin, dimitron, folcodal, midronal, mitronal, stutgeron, glanil
CHEBI:31403
C1CN(CCN1CC=CC2=CC=CC=C2)C(C3=CC=CC=C3)C4=CC=CC=C4

Specifications

298-57-7
MFCD00056037
cinnarizine, stugeron, dimitronal, stutgin, dimitron, folcodal, midronal, mitronal, stutgeron, glanil
DERZBLKQOCDDDZ-JLHYYAGUSA-N
1-benzhydryl-4-[(E)-3-phenylprop-2-enyl]piperazine
1547484
368.52
C26H28N2
14,2305
Soluble in chloroform at 50mg/ml
C1CN(CCN1CC=CC2=CC=CC=C2)C(C3=CC=CC=C3)C4=CC=CC=C4
368.524
CHEBI:31403
Cinnarizine
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 206-064-8

missing translation for 'rtecsNumber' : TL3430000

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.