missing translation for 'onlineSavingsMsg'
Learn More

Cinnarizine, MedChemExpress

Product Code. 16418606
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16418606 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16418606 Supplier Medchem Express Supplier No. HYB1090
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Cinnarizine is an antihistamine and a calcium channel blocker, promote cerebral blood flow, used to treat cerebral apoplexy, post-trauma cerebral symptoms, and cerebral arteriosclerosis.

Cinnarizine is an antihistamine and a calcium channel blocker, promote cerebral blood flow, used to treat cerebral apoplexy, post-trauma cerebral symptoms, and cerebral arteriosclerosis.

Specifications

Chemical Name or Material Cinnarizine
CAS 298-57-7
Color White
Purity Grade Notes Research
Molecular Formula C26H28N2
Quantity 100 mg
Solubility Information DMSO : 7.14 mg/mL (19.38 mM; Need ultrasonic)
SMILES N1(C(C2=CC=CC=C2)C3=CC=CC=C3)CCN(C/C=C/C4=CC=CC=C4)CC1
Molecular Weight (g/mol) 368.51
Formula Weight 368.51
Percent Purity 99.63%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.