Learn More
Cinnamylamine Hydrochloride, TRC
Product Code. 30368309
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30368309 1 g 1g
30349212 100 mg 100mg
30351080 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30368309 Supplier Toronto Research Chemicals Supplier No. TRCC4420901G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5586-89-0
Molecular Formula C9 H11 N . Cl H
Molecular Weight (g/mol) 169.65
Synonym 3-Phenyl-2-Propen-1-amine Hydrochloride (1:1), 2-Propen-1-amine, 3-phenyl-, hydrochloride (9CI), Cinnamylamine, hydrochloride (6CI,7CI,8CI)
IUPAC Name (E)-3-phenylprop-2-en-1-amine;hydrochloride
SMILES Cl.NC\C=C\c1ccccc1
Specifications
Quantity 1 g
InChI Formula InChI=1S/C9H11N.ClH/c10-8-4-7-9-5-2-1-3-6-9;/h1-7H,8,10H2;1H/b7-4+;
Formula Weight 169.0658
Chemical Name or Material Cinnamylamine Hydrochloride