Learn More
Cinnamoyl Chloride, Sigma-Aldrich™
Specifications
Specifications
| CAS | 17082-09-6 |
| Melting Point | 32°C to 36°C |
| Density | 1.16 g/cm3 at 20°C |
| Molecular Formula | C6H5CH=CHCOCl |
| MDL Number | MFCD00000986 |
| Quantity | 100 mL |
| Synonym | Cinnamoyl chloride, Cinnamic acid chloride, Cinnamoyl chloride |
| Molecular Weight (g/mol) | 166.6 |
| Grade | Research |