missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Ciclopirox
GREENER_CHOICE

Product Code. 18771108
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18771108

Brand: Tocris Bioscience™ 6384/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Pan-histone demethylase inhibitor

Ciclopirox is a pan-histone demethylase inhibitor. Exhibits no significant effect on histone deacetylases. Inhibits MYC-signaling and proliferation of neuroblastoma cells. Promotes Cdc25A-degradation in cancer cells. Antifungal.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6-Cyclohexyl-1-hydroxy-4-methyl-2(1H)-pyridinone
CAS 29342-05-0
Assay Percent Range >99%
Quantity 10 mg
Molecular Formula C12H17NO2
Formula Weight Observed MW: 207.27
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.