Learn More
CI-1040, MedChemExpress
Product Code. 16478068
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
25mg
50mg
5mg
Product Code. 16478068
Brand: Medchem Express HY50295
MedChemExpress CI-1040 (PD 184352) is an orally active, highly specific, small-molecule inhibitor of MEK with an IC50 of 17 nM for MEK1.
CI-1040 (PD 184352) is an orally active, highly specific, small-molecule inhibitor of MEK with an IC50 of 17 nM for MEK1.
Specifications
Chemical Name or Material CI-1040
CAS 212631-79-3
Color White
Purity Grade Notes Research
Molecular Formula C17H14ClF2IN2O2
Quantity 10 mg
Synonym PD 184352
Solubility Information DMSO : ≥ 100 mg/mL (208.92 mM)
SMILES O=C(C1=CC=C(C(F)=C1NC2=CC=C(I)C=C2Cl)F)NOCC3CC3
Molecular Weight (g/mol) 478.66
Formula Weight 478.66
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease