missing translation for 'onlineSavingsMsg'
Learn More

α-Chymotrypsin, MP Biomedicals™

Product Code. 11496105 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11496105 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11496105 Supplier MP Biomedicals Supplier No. 0215227210
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Preferentially catalyzes peptide bonds involving L-isomers of tyrosine, phenylalanine, and tryptophan. α-Chymotrypsin, MP Biomedicals is a serine protease that hydrolyzes peptide bonds with aromatic or large hydrophobic side chains on the carboxyl e

α-Chymotrypsin readily acts upon amides and esters of susceptible amino acids. In addition to bonds involving aromatic amino acids, it catalyzes at a high rate the hydrolysis of bonds of leucyl, methionyl, asparaginyl, and glutamyl residues.

TRUSTED_SUSTAINABILITY

Specifications

CAS 9005-12-3
Molecular Formula C7H8OSi
Quantity 10 g
Synonym methylphenylsilanone, polyphenylmethylsiloxane, methylphenyl oxo silane, silane, methyloxophenyl, methyl phenyl silanone
InChI Key IXKNJYSAWNJQQY-UHFFFAOYSA-N
SMILES C[Si](=O)C1=CC=CC=C1
IUPAC Name methyl-oxo-phenylsilane
Molecular Weight (g/mol) 136.23
PubChem CID 3032548
Formula Weight 25 kDa (Lit.)
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.