Learn More
Proteintech ChromoTek TurboGFP-Trap Magnetic Agarose Kit
Description
- TurboGFP-Trap Magnetic Agarose is an affinity reagent for immunoprecipitation (IP) of TurboGFP fusion proteins
- Consists of an anti-TurboGFP VHH/ Nanobody that is coupled to magnetic agarose beads
Specifications
Specifications
| Quantity | 20 Reactions |
| Sufficient For | 20 Reactions |