Learn More
Proteintech ChromoTek Nano-CaptureLigand™ Mouse IgG2b, Fc-specific VHH, Biotinylated
Product Code. 17313493
Change view
Click to view available options
Quantity:
10 μL
100 μL
Unit Size:
100µL
10µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17313493 10 μL 10µL
17323493 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17313493 Supplier Proteintech Supplier No. smsG2bB110
Nano-CaptureLigand for mouse IgG2b: Fc-specific single domain antibody/VHH [CTK0106], biotinylated.
  • The Nano-CaptureLigand mouse IgG2b captures and immobilizes mouse IgG2b antibodies and Fc-fragments to avidin/streptavidin coated surfaces used in BLI, SPR, and ELISA assays
  • Consists of a biotinylated Nanobody that specifically binds to mouse IgG2b antibodies with high affinity and in a site-directed manner
Specifications
Quantity 10 μL
Target Specificity Fc-fragment of Mouse IgG2b
Conjugate Biotin
Clonality Monoclonal
Content And Storage Upon receipt store at +4°C/+40°F. Optional: Aliquot upon arrival and store at -20°C/-4°F
For Use With (Application) BLI, SPR, ELISA