Learn More
Proteintech ChromoTek Nano-CaptureLigand™ Human IgE, Recombinant VHH, Biotinylated
Product Code. 17333463
Change view
Click to view available options
Quantity:
10 μL
100 μL
Unit Size:
100µL
10µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17333463 10 μL 10µL
17343463 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17333463 Supplier Proteintech Supplier No. shuEB110
Nano-CaptureLigand for human IgE: single domain antibody/VHH [CTK0111], biotinylated.
  • The ChromoTek Nano-CaptureLigand™ human IgE, VHH, biotinylated is used for the site-directed and specific immobilization of human IgE in biosensor and ELISA assays
  • It captures non-biotinylated human IgE antibodies to streptavidin/avidin
  • Nano-CaptureLigand human IgE, VHH, biotinylated comprises a monoclonal biotinylated VHH/ Nanobody
Specifications
Quantity 10 μL
Target Specificity Fc-fragment of Human IgG1, IgG2, IgG3, and IgG4, Fc-fragment of Rabbit IgG
Conjugate Biotin
Clonality Monoclonal
Content And Storage Upon receipt store at +4°C/+40°F. Optional: Aliquot upon arrival and store at -20°C/-4°F
For Use With (Application) BLI, SPR, ELISA