Learn More
Proteintech ChromoTek Histone-Chromobody™ Plasmid (eGFP)
Product Code. 17363523
Change view
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17363523 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17363523 Supplier Proteintech Supplier No. tcg
Histone-Chromobody™ fused to EGFP.
  • DNA plasmid encoding for anti-H2A-H2B VHH (anti-H2A-H2B Nanobody) fused to EGFP
Specifications
Quantity 20 μg