Learn More
Proteintech ChromoTek Halo-Trap Magnetic Agarose Kit
Description
- Halo-Trap Magnetic Agarose Kit is used to immunoprecipitate Halo-tagged fusion proteins from cell extracts of various organisms like mammals, plants, bacteria, yeast, insects etc. in the absence or presence of a covalently bound ligand
- The interaction between Halo-Trap and the Halo-tagged fusion protein is reversible
Specifications
Specifications
| Quantity | 20 Reactions |
| Sufficient For | 20 Reactions |