missing translation for 'onlineSavingsMsg'
Learn More

Proteintech ChromoTek GFP-Booster Alexa Fluor™ 568

Product Code. p-7235978
Click to view available options
Quantity:
10 μL
50 μL
Unit Size:
10µL
50µL
This item is not returnable. View return policy

Product Code. 17363333

Brand: Proteintech gb2AF56810

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

GFP VHH conjugated to Alexa Fluor™ 568.

  • anti-GFP VHH/ Nanobody conjugated to a fluorescent dye for IF/ microscopy

Specifications

Clonality Monoclonal
Quantity 10 μL
For Use With (Application) Immunofluorescence (IF), Immunohistochemistry (IHC), Immunocytochemistry (ICC)
Conjugate Alexa Fluor™ 568
Target Specificity AcGFP, Citrine, CFP, eGFP, eYFP, GFP S65T, pHluorin, sfGFP, mClover (Clover A206K), TagGFP, tagGFP2, Venus, wtGFP, YFP
Content And Storage Shipped at ambient temperature. Store at -20°C/-4°F. Avoid freeze-thaw cycles. Aliquot upon arrival. Protect from light. Stable for 6 months.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.