Learn More
Chromium(0) Hexacarbonyl, Sigma-Aldrich™
Product Code. 15894661
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15894661 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
MQ200 Graded - For Research and Non-regulated Industrial Applications
Product Code. 15894661 Supplier Sigma Aldrich Supplier No. 8221960025
 Restricted Item
Estimated Shipment: 20-11-2026
MQ200 Graded - For Research and Non-regulated Industrial Applications
For synthesis
TRUSTED_SUSTAINABILITY
Specifications
CAS 13007-92-6
Melting Point 149°C to 150°C
Density 1.77 g/cm3 at 20°C
Molecular Formula Cr(CO)6
MDL Number MFCD00010945
Quantity 25 g
Synonym Chromium(0) hexacarbonyl, Chromiumcarbonyl, Chromium(0) hexacarbonyl
Molecular Weight (g/mol) 220.06
Grade Research