Learn More
Macherey-Nagel™ Chrombond™ RS 4 SiOH Polypropylene Flash Column
Product Code. 12777740 Shop All Macherey-Nagel Products
Change view
Click to view available options
I.D.: 12.4mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12777740 I.D.: 12.4mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12777740 Supplier Macherey-Nagel™ Supplier No. 732800
Save Up to 60% on Contract Testing Essentials
Use promo code 28025
View all promotions
In Stock
Specifications
Length (Metric) 98 mm
Material Polypropylene
Type Flash Column
Column Type Flash Column
For Use With (Equipment) Directly Used in The Teledyne Isco Companion, Rf