Learn More
Macherey-Nagel™ Chrombond™ RS 330 C18 ec Polypropylene Flash Column
Product Code. 12758332 Shop All Macherey-Nagel Products
Change view
Click to view available options
Length (Metric):
270 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
12758332 270 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12758332 Supplier Macherey-Nagel™ Supplier No. 732817
Save Up to 60% on Contract Testing Essentials
Use promo code 28025
View all promotions
In Stock
Specifications
Length (Metric) 270 mm
Material Polypropylene
Type Flash Column
Column Type Flash Column
For Use With (Equipment) Directly Used in The Teledyne Isco Companion, Rf
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.