Learn More
Macherey-Nagel™ Chrombond™ RS 15 SiOH Polypropylene Flash Column
Product Code. 12787740 Shop All Macherey-Nagel Products
Change view
Click to view available options
Length (Metric):
116 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
12787740 116 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12787740 Supplier Macherey-Nagel™ Supplier No. 732801
Save Up to 60% on Contract Testing Essentials
Use promo code 28025
View all promotions
In Stock
Ideal for Flash separations from 10 mg up to 160 g. Macherey Nagel™ Chrombond™ RS 15 SiOH Polypropylene Flash Column provides convenient operation and reliable upscaling
  • Increases flexibility considerable program of different phases and modifications
  • Saves time and money convenient prices, short delivery times
  • Increases analytical safety high pressure stability up to 21 bar / 300 psi (21 bar for cartridges < 40 g, 15 bar for 80 g and 120 g cartridges, 12 bar for cartridges > 200 g) excellent separation efficiency, good reproducibility
Specifications
Length (Metric) 116 mm
Material Polypropylene
Type Flash Column
Column Type Flash Column
For Use With (Equipment) Directly Used in The Teledyne Isco Companion, Rf