Learn More
Cole Parmer Chromatography Research Supplies PURESEP-T™ Septa
Product Code. 11844401 Shop All Cole Parmer Products
Change view
Click to view available options
Quantity:
50/Pk.
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11844401 50/Pk. Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11844401 Supplier Cole Parmer Supplier No. 0199420
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Chromatography Research Supplies PURESEP-T™ Septa are made of PTFE and polyimide films on silicone rubber bases. PTFE gives maximum inertness and low bleed
Specifications
Product Type Low Bleed Septum
Temperature (English) Max. 437°F
Temperature (Metric) Max 225°C
Material Silicone with PTFE and Polyimide Films
Diameter (Metric) Inner 6 mm
Quantity 50/Pk.