Specifications
| Base Material | Silica |
| Description | Adsorbent Weight: 450 mg |
| For Use With (Application) | Amino Acids, Amines, Chlorophyll, PCBs |
| Particle Size | 45 μm |
| Sorbent | SA |
| Type | Chromafix SA Cartridge |
| Pore Size | 60 Å |
| Quantity | 50 Pack |
| Surface Area (Metric) | 500 m2/g |