Learn More
Macherey-Nagel™ Chromafix™ PS-H+ SPE Cartridge
Product Code. 10125091 Shop All Macherey-Nagel Products
Change view
Click to view available options
Bed Weight:
230 g
900 g
Unit Size:
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Bed Weight unitSize
10125091 230 g Pack of 50
12471718 900 g Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10125091 Supplier Macherey-Nagel™ Supplier No. 731867
In Stock
Add to basket
Add to basket
For removal or concentration of cations from water, decreasing the pH value of basic samples for standard protocol. Macherey-Nagel™ Chromafix™ PS-H+ SPE Cartridge is a polymer-based SPE phase for RP and ion chromatography.
These polymer SPE phases are used for the removal of interfering compounds and enrichment of the analytes in reverse phase and ion chromatography.
  • Strong PS/DVB cation exchanger, with H+ form capacity of 2.9 meq/g
  • Very low degree of swelling, thus very well suited for chromatography
  • Reliable function over the whole pH range from 0–14
  • Improves chromatographic separation if the interfering components overlap with the analyte
  • Improves lifetime of the column, since interfering components can block the packing
Specifications
Base Material Strong PS/DVB Cation Exchanger, H+ form
Bed Weight 230 g
For Use With (Application) Removal or Concentration of Cations From Water Decreasing The pH Value of Basic Samples
Particle Size 100 μm
Type Chromafix PS-H+ Cartridge
Pore Size 100 Å
Quantity 50 Pack
Tube Size S